Would you like to log in again?
Phenol Red Sucrose Broth
Supplier: VWR International
AD-MM1037-500G AD-MM1037-100G
470317-436EA 136.1 USD InStock
470317-436 470317-434
Phenol Red Sucrose Broth
Media for Microbiology Dehydrated Media for Microbiology
A basal medium containing a single carbohydrate used to determine whether bacteria can ferment the specific carbohydrate. The medium contains a pH indicator that changes from red to yellow if the bacteria can ferment the carbohydrate.
  • May be used with Durham tubes to determine if gas is produced during the fermentation process
To prepare media, add 20 g of phenol red lactose broth to a liter of distilled or de-ionized water. The media will not be sterile until autoclaved prior to use.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty