Would you like to log in again?
DURAN® Tamper Evident Red/Blue Screw Cap GL 45 with Linerless Plug Seal, Qorpak®
Supplier: Qorpak
DURAN®
289201
76308-364CS 60.67 USD InStock
76308-364
DURAN® Tamper Evident Red/Blue Screw Cap GL 45 with Linerless Plug Seal, Qorpak®
Closures Bottle Closures
Seal GL 45 Media Bottles with secure cap.
  • Tamper evident
  • Autoclavable
  • Linerless Plug Seal
Polypropylene material allows maximum temperature of 140 °C with minimum temperature of –40 °C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty