Would you like to log in again?
Cardionics® SAM 3G- Student Auscultation Manikin
Supplier: AMERICAN 3B SCIENTIFIC SE
8000938 1022396 1021557 8000939 1021554
470317-912EA 3826.79 USD InStock
470317-912 470318-038 470317-910 470317-952 470317-908
Cardionics® SAM 3G- Student Auscultation Manikin
Models Human Biology Models
An advanced auscultation learning package.
  • Includes COVID sounds
  • Lightweight and portable
  • User friendly software
  • Phonocardiogram
  • Large library of sounds videos
  • Complete lesson guide
  • Use with any stethoscope
  • Palpable carotid pulse
  • Create and save case videos
Auscultation is an essential clinical skill needed to assess and monitor patients’ conditions. With the new Cardionics SAM 3G Student Auscultation Manikin 3rd Generation, teaching and learning this skill becomes even more versatile and economic. SAM 3G Trainer can be used with any stethoscope available and offers the largest Real Sound Library, Case Videos, Echo Cardiogram Videos and ECG Waveforms for students to master identification of various heart, lung and bowel sounds. All sounds and videos are recorded from live patients for students to experience a life-like simulation. Case videos contain patient and clinical interactions for various conditions and enhance the learning by providing visual representation of the sounds. Users can also create and save their own case videos.
SAM 3G Student Auscultation Manikin is world’s most innovative product available on the market, used in teaching and learning heart, lung and bowel sounds. When connected to the laptop (included) with the pre-installed software, a variety of sounds, videos and lessons recorded can be accessed.
Additionally, when connected to speakers SAM 3G Student Auscultation Manikin can be easily moved into a classroom or an auditorium for group instruction, where a larger audience of students can benefit from a co-learning experience and share their thoughts at the same time as high quality heart, lung and bowel sounds are emitted. The programmable and password protected lectures allow teaching institutions to have numerous instructors utilize the SAM 3G Trainer. Included with the product is SAM’s Lesson Guide: a complete guide of lessons for each auscultation sound that SAM 3G offers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty