Would you like to log in again?
3B Scientific® Hall Effect Apparatus
Supplier: 3B Scientific
3B Scientific®
1008522 1009934
470316-690EA 1087.09 USD InStock
470316-690 470316-692
3B Scientific® Hall Effect Apparatus
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Basic apparatus for providing contact, power supply and support to a germanium crystal on a circuit board in experiments on the Hall effect or on conductivity. Includes an integrated, adjustable constant current source to provide the current through the sample, a measuring amplifier with offset compensation for Hall voltages and heating to raise the crystal to as high as 170 °C, also featuring temperature regulation and a switchable display showing Hall voltage, sample current, sample voltage or temperature.
Hall voltage and sample voltage can be tapped directly from the front panel. In addition three equivalent voltage outputs for Hall voltage, sample current and sample temperature can be measured from the side. Includes an attachment for assembling the apparatus on the U-shaped core of a transformer assembly kit and two connecting leads with eight-pin miniDIN plugs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty