Would you like to log in again?
BTX Mini Micro-Grabber Electrode Connection Cables, Harvard Apparatus
Supplier: Harvard Apparatus
BTX
45-0503
95062-356EA 98.42 USD InStock
95062-356
BTX Mini Micro-Grabber Electrode Connection Cables, Harvard Apparatus
Cables
Mini micrograbber cables are an ideal to connect specialty in vivo and in virtro electrodes to electroporation generators.
  • Retractable micrograbber tips
  • Compatible with BTX generators
  • Color coded red and black cables
  • Cable set provides easy electrical polarity identification at a glance
Mini micrograbber cables offer an easy-to-use design with BTX connection type. Spring loaded micrograbber tips provide secure attachment to electrodes on one end of the cables, and 2 mm diameter pin tips on the other end of the cables attach to Tweezertrode adapter cables for connection to BTX electroporators that accept 4 mm banana cables.
Mini micrograbber cables are recommended for use with the following BTX electrodes: Genetrodes, Gene Paddles, Oocyte Electrode, Oocyte Petri Electrode and Petri Dish Patinum Electrode Chambers for Tissues.
Generator compatibility: Gemini X2, ECM 830, Legacy ECM 2001, ECM 2001+.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty