Would you like to log in again?
Corepoint Scientific™ General Purpose Laboratory Refrigerator, Horizon Scientific
Supplier: Horizon Scientific
LR301WWW/0
76333-618EA 9380.81 USD InStock
76333-618
Corepoint Scientific™ General Purpose Laboratory Refrigerator, Horizon Scientific
Refrigerators
These auto defrost laboratory freezers offer multi-purpose cold storage for scientific and industrial settings.
  • Mechanical thermostats
  • Keyed door locks
  • Interior shelving
  • Probe access port
These general purpose refrigerators provide multi-functional features in a cost-effective design.
Ordering information: Units come with a two year parts and labor warranty, plus an additional three year compressor parts warranty.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty