Would you like to log in again?
Corepoint Scientific™ Premier™ Hydrocarbon Manual Defrost Solid Door Freezer, Horizon Scientific
Supplier: Horizon Scientific
Corepoint Scientific™ Premier™
NSPF141WWW/0MHC
76333-622EA 3450.99 USD InStock
76333-622
Corepoint Scientific™ Premier™ Hydrocarbon Manual Defrost Solid Door Freezer, Horizon Scientific
Freezers Laboratory / General Purpose Freezers
These efficient manual defrost freezers provide premier cooling for laboratory settings. They offer storage solutions for a variety of scientific and clinical applications.
  • Microprocessor temperature control
  • Integrated temperature alarms
  • Keyed door locks
  • Probe access
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
Units come equipped with forced draft circulation and precise temperature monitoring through microprocessor control. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Certifications: *Select models ENERGY STAR certified: 76333-622
Ordering information: All units come with a two year parts and labor warranty, plus an additional three year compressor parts warranty.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty