Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eosin Y (yellowish) in aqueous solution, HARLECO® counterstain in H&E, for histology, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: Tetrabromofluorescein disodium saltEosin Y disodium saltEosin 2252′,4′,5′,7′-Tetrabromofluorescein disodium saltSolvent Red 43Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoateDisodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate)Field's Stain B2,4,5,7-Tetrabromofluorescein disodium salt

592-75 592-75
15204-258EA 156.46 USD InStock
15204-258 CA15204-258L
Eosin Y (yellowish) in aqueous solution, HARLECO® counterstain in H&E, for histology, Sigma-Aldrich®
Eosin Y (yellowish)
HARLECO® cytoplasmic counterstain for H&E.

  • Prepared from dyes certified by the Biological Stain Commission (BSC)
  • For In-Vitro Diagnostic Use
  • For Professional Use


Formula: C₂₀H₆Br₄Na₂O₅
MW: 691.86 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005040
CAS Number: 17372-87-1
IVDLogo
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty