Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Alcohol 100%, HARLECO® for histology, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: Reagent alcohol

65347-85
34172-020CS 291.83 USD InStock
34172-020
Alcohol 100%, HARLECO® for histology, Sigma-Aldrich®
Alcohol
Part of the QSM Histology stain line. Used to hydrate or dehydrate tissue samples in tissue processing and staining protocols.


Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1987
ADR: 3,II
Order Now
Specification Test Results
Expiration date 18 months from mfg date
Assay (Ethanol) 89.0-92.0%
Assay (Isopropyl alcohol) 4.0-6.0%
Assay (Methanol) 3.5-5.5%
Color (APHA) 10APHA max
Form Clear liquid
Infrared Spectrum Conforms to standard
Residue after evaporation 0.001% max
Water (H2O) 0.075% max

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty