Would you like to log in again?
Acroprep™ Filter Plates with GHP Membrane, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
AcroPrep™ Advance
8282
89233-866PK 193.89 USD InStock
89233-866
Acroprep™ Filter Plates with GHP Membrane, Cytiva (Formerly Pall Lab)
Microplates Filter Plates
Pall GHP Polypropylene membrane will be discontinued effective October 31st, 2019. This discontinuation affects devices including Filter Plates, Syringe Filters, Centrifugal Devices and Membrane Filter Discs.
  • Download our application note under "Related Items" below to see how our next generation universal membrane wwPTFE outperforms the GHP membrane
The GHP membrane used in Pall Laboratory Acrodisc® syringe filters, centrifugal devices and Acroprep™ filter plates will be discontinued due to the rationalization of a key material from our supplier.
However we are excited to announce the launch of the NEW wwPTFE universal membrane as a direct replacement for GHP filtration products. Pall’s next generation of universally chemically compatible membrane, our water wettable polytetrafluoroethylene (wwPTFE) membrane, yielded significant performance advantages compared to GHP when extractables, binding, column life and chemical compatibility were evaluated.
With the NEW wwPTFE Universal Membrane:
• Extend column life up to 52 times
• 24% less extractable materials than competitive filters
• 10x lower extractable materials than hydrophilic polypropylene
• Less API binding than filters with hydrophilic polypropylene
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty