Would you like to log in again?
Scotch® 3750 Commercial Grade Packaging Tape, 3M
Supplier: 3M
MCO 3750260CR
82003-546EA 20.57 USD InStock
82003-546
Scotch® 3750 Commercial Grade Packaging Tape, 3M
Tapes
Scotch® Commercial-Grade Shipping and Packaging Tape is designed for commercial performance.
  • Time-proven construction assures superior performance
  • Excellent holding power for heavy-duty packaging and shipping
  • Designed to keep box seams closed during rough handling
  • Dispenser not included
  • Hot melt adhesive material
This tape combines strong, hot-melt adhesive and a sturdy backing to seal out moisture, dust and dirt. Resists splitting and ensures reliable closures.
Certifications: Meets U.S. Postal Regulations.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty