Would you like to log in again?
Squishy Circuits, Dough Kit
Supplier: SQUISHY CIRCUITS STORE LLC SE
DOUGH
470317-846EA 22 USD InStock
470317-846
Squishy Circuits, Dough Kit
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Learn and explore through the power of play. With Squishy Circuits dough kit, make play dough science projects fun through an array of different colors.
  • Invent
  • Create
  • Explore
The dough does contain gluten, but is safe and non-toxic. The conductive dough does dry out if left out of the container, but the insulating dough does not dry out.
Delivery information: Kit includes: 7 containers of conductive dough and 1 container of white insulating dough, Each container holds 7 ounces (a normal, bought container is 5 ounces).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty