Would you like to log in again?
Filter Adapter Cap, TriForest Enterprises
Supplier: TriForest
FAN-38
10014-832EA 48.49 USD InStock
10014-832
Filter Adapter Cap, TriForest Enterprises
Closures Safety Caps and Safety Waste Caps
TriForest has designed a polypropylene filter adapter to combine the filtration options of preferred 38-430 bottleneck filters with a rugged labware product line.
  • Autoclavable
  • Easy to use and store
  • Reusable
The filter adapter cap provides a leakproof alternative that increases product utility and media containment.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty