Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gas Manifolds, 4-Place, Glass Stopcocks, Chemglass

Supplier: Chemglass
CG-1814-01
80071-218EA 732.29 USD InStock
80071-218
Gas Manifolds, 4-Place, Glass Stopcocks, Chemglass
Manifolds
Four port manifold with 2 mm bore glass stopcocks on the ports.

  • A 'T' bore stopcock is located 100 mm from the end of the manifold and 40 mm from the first port
  • The distance between the other three ports is 50 mm
  • The last port is 75 mm from the opposite end of the manifold

The entire manifold is constructed of capillary tubing having an O.D. of 8 mm and an I.D. of 2 mm.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty