Would you like to log in again?
Distilling Receivers, Chemglass
Supplier: Chemglass
CG-1261-A-12 CG-1261-A-49 CG-1261-A-10
16002-694EA 415.4 USD InStock
16002-694 16002-704 16002-692
Distilling Receivers, Chemglass
Distillation Components Distillation Receivers
Graduated distilling receivers with a standard taper outer joint on top.
  • Stopcocks are either a 2 or 4 mm PTFE
  • Distance between the receiver and the uptake arm: 10 and 20 ml are 100 mm center to center
  • The 50, 100 and 250 ml are 150 mm center to center
They have standard taper lower drip joint.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty