Would you like to log in again?
Rotary Evaporator Bump Trap, Self-Washing, Chemglass
Supplier: Chemglass
CG-1319-05
80068-056EA 271.15 USD InStock
80068-056
Rotary Evaporator Bump Trap, Self-Washing, Chemglass
Vacuum Traps
Trap is designed to eliminate hold up common to the round style of rotary evaporator traps.
  • Tapered sidewalls permit continuous washing with condensed solvent
  • While preventing vapor cooling points along the path of the condenser
The upper stem has four large holes flush with the lower end allowing solvent vapor to flow freely and making this portion of the trap easy to clean. The increased volume of the trap prevents 'bumping' and keeps solvent out of the vapor tube.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty