Would you like to log in again?
Condensers, Reflux, Double Wall Thimble Shape Bulb, Chemglass
Supplier: Chemglass
CG-1217-25
16002-680EA 904.76 USD InStock
16002-680
Condensers, Reflux, Double Wall Thimble Shape Bulb, Chemglass
Condensers
Compact high efficiency reflux condensers feature a double wall thimble shape bulb sealed into the outer jacket.
  • Efficiency of a standard 400 mm Liebig or West condenser
Serrated hose connections for water connection extend through the outer bulb and have an O.D. of 10 mm at the middle serration.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty