Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Picric acid 0,1% in acetone

Supplier: Electron Microscopy Sciences
Danger

19553
101410-762EA 93.33 USD InStock
101410-762
Picric acid 0,1% in acetone
Picric acid


Formula: C₆H₃N₃O₇
MW: 229.11 g/mol
MDL Number: MFCD00007102
CAS Number: 88-89-1
UN: 1090
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/48/77/28984877.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty