Would you like to log in again?
Rotary Evaporator Bump Traps, Elliptical, Chemglass
Supplier: Chemglass
CG-1320-E-10
89166-576EA 222.02 USD InStock
89166-576
Rotary Evaporator Bump Traps, Elliptical, Chemglass
Vacuum Traps
Evaporator bump trap prevents contents of flask from being drawn into the condenser.
  • This allows the use of larger recovery flasks in a rotary evaporator
  • Traps have a 24/40 top outer joint and the listed size of lower inner joint
Rotary evaporator bump trap is used between the vapor tube and evaporation flask on a rotary evaporator to prevent contents of the flask from being drawn into the condenser in the event of 'bumping'. Unique elliptical shape provides large trap volume while decreasing the overall length when compared to standard spherical designs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty