Would you like to log in again?
Condensers, Double Coils, Rotary Evaporators, Chemglass
Supplier: Chemglass
CG-1326-10
80060-928EA 1487.91 USD InStock
80060-928
Condensers, Double Coils, Rotary Evaporators, Chemglass
Condensers
Double coil Condenser for use with rotary evaporators for cooling tap water in a diagonal position.
  • Condenser has a 50 mm O.D. flange at the bottom, a 19/38 top valve
  • Removable hose connections using GL-14 glass threads
Delivery information: Condenser is supplied complete with plastic caps and serrated hose connections.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty