Would you like to log in again?
Condensers, Double Coils, B Assembly, for Rotary Evaporators, Plastic Coated, Chemglass
Supplier: Chemglass
CG-1329-10
80060-948EA 1223.31 USD InStock
80060-948
Condensers, Double Coils, B Assembly, for Rotary Evaporators, Plastic Coated, Chemglass
Condensers
Double coil condenser for use with rotary evaporators.
  • For cooling with tap water in a vertical position
Condenser has a 45/40 lower inner drip tip joint, a 29/32 top outer joint, and removable hose connections using GL 14 glass threads.
Delivery information: Condenser is supplied complete with plastic caps and serrated hose connections.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty