Would you like to log in again?
Condensers, Allihn, Static Waterless, Chemglass
Supplier: Chemglass
CG-1206-W214 CG-1206-W224
76299-876EA 619.65 USD InStock
76299-876 76299-874
Condensers, Allihn, Static Waterless, Chemglass
Condensers
Static waterless Allihn style reflux condenser with a standard taper outer joint at the top and a lower inner drip tip joint and GPI thread on side for filling the condenser with ethylene glycol.
  • No issues with water leakage or potential flooding
  • By not using water the environmental impact is reduced as is the costs of water usage
  • Clear visibility of the reaction reflux
  • Suitable for use under vacuum
  • For solvent volumes up to one liter
  • Waterless allihn condenser is considerably cost effective than other waterless condenser options
A standard taper joint at the top and a lower inner drip tip joint and GPI thread on the side for filling the condenser with ethylene glycol.
Ordering information: Supplied with open top cap and septa for closing off once the condenser is filled (Condenser is not supplied with ethylene glycol).
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty