Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Alcian blue solution pH 2.5

Supplier: Leica Biosystems
Danger
Synonyms: C.I. 74240C.N. AnAn-6Ingrain Blue 1Alcian blue 8 GX

Surgipath®
38016SS3A
75810-392EA 395.62 USD InStock
75810-392
Alcian blue solution pH 2.5
Alcian blue 8 GX
Component of the Alcian Blue Stain Kits.

Alcian Blue is normally prepared at an acidic pH of 2.5.


Formula: C₅₆H₆₈Cl₄CuN₁₆S₄
MW: 1298.86 g/mol
MDL Number: MFCD00010720
CAS Number: 33864-99-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty