Would you like to log in again?
Distillation Receiver, Graduated, Chemglass
Supplier: Chemglass
CG-1277-14
80067-902EA 444.62 USD InStock
80067-902
Distillation Receiver, Graduated, Chemglass
Distillation Components Distillation Receivers
Cow type receiver has a single standard taper outer joint at the top.
  • Four lower graduated receiver tubes
  • Can be accessed via a syringe for product recovery
Tubes are graduated from 0 to 3ml in 0.2 ml subdivisions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty