Would you like to log in again?
Distilling Heads, Vigreux, Micro, Chemglass
Supplier: Chemglass
CG-1241-05
80067-626EA 669.47 USD InStock
80067-626
Distilling Heads, Vigreux, Micro, Chemglass
Distillation Components Distillation Heads
Distillation heads feature immersion thermometer.
  • Distillation head with condenser
  • Sealed to a vigreux column
Top thermometer joint is a 10/18 for use with a 1" immersion thermometer. Vacuum adapter has an outer joint and accepts a 80067-922 distribution adapter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty