Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Orange G 0,5% in aqueous solution

Supplier: Electron Microscopy Sciences
Synonyms: Disodium 7-hydroxy-8-phenylazonaphthalene-1,3-disulphonateAcid Orange 10Wool Orange 2GOrange G sodium saltCI 16230

26066-05
101410-940EA 110.17 USD InStock
101410-940
Orange G 0,5% in aqueous solution
Orange G


Formula: C₁₆H₁₀N₂Na₂O₇S₂
MW: 452.38 g/mol
MDL Number: MFCD00012457
CAS Number: 1936-15-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/49/26/28984926.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty