Would you like to log in again?
Test Tube pH Pen, Penetration, Sper Scientific
Supplier: Sper Scientific
850067
76333-912EA 238.77 USD InStock
76333-912
Test Tube pH Pen, Penetration, Sper Scientific
Electrochemical Meters pH and Multi-Parameter Meters
Fits standard test tubes 16×50 mm or larger.
  • Penetration probe for food and soil
  • Highly stable and accurate readings with a large LCD display
  • Designed for simple one-hand operation
  • Features hold function, and auto power off with low battery indicator
Automatically calibrates to pH 4, 7 and 10 buffers and can be adjusted to recognize some other non-standard buffers as well.
The case is IP 65 waterproof and will float if accidentally dropped into the water.
Delivery information: Comes with two button-cell batteries and a built-in electrode.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty