Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
F.I.T.™ Comfort Deck Open, Wearwell®
Supplier: WEARWELL
F.I.T.™
548.58X12X12BK-CS20 715411001910
76048-790CS 199.52 USD InStock
76048-790 76048-788
F.I.T.™ Comfort Deck Open, Wearwell®
Floor Mats Traction and Anti-Fatigue Floor Mats
Most versatile modular tile.
  • Customizable in 3" increments
  • Secure and seamless connections
  • Ability to replace single tiles saves money and down time
  • Silicone-free
The most versatile 12×12" interlocking ergonomic flooring system in the market. The patented LockSafe® connections simply snap together and configure to fit your exact workspace. The top surface and underside of F.I.T. tiles are designed to allow rapid drainage of fluids.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty