Would you like to log in again?
Condensers, Reflux, Removable Hose Connections, Chemglass
Supplier: Chemglass
CG-1213-A-HC01
47733-782EA 562.51 USD InStock
47733-782
Condensers, Reflux, Removable Hose Connections, Chemglass
Condensers
Reflux Condenser similar to CG-1213-A but with detachable hose connections.
  • Connections are made of polypropylene and provide a safe and easy way of connecting and disconnecting flexible tubing to the condenser
Hose connections have an O.D. of 8 mm and an I.D. of 4 mm.
Ordering information: Supplied complete with two GL-14 open top screw caps. See CG-198 and CG-195 for replacement GL fittings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty