Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Upright Microscopes, BA410 Elite (Motic)
Supplier: Motic
Motic
1100100403783 1100100403881
470321-016EA 1538.99 USD InStock
470321-016 470321-014
Upright Microscopes, BA410 Elite (Motic)
Microscopes
The BA410 Elite is designed for diagnosis in pathology, hematology and cytology.
  • Increased numerical apertures for detailed specimen evaluation
  • Energy saving mode: Automatic On/Off setting via infrared sensor
  • Halogen light interchangeable with a LED module (to be purchased separately) for the 50 W version
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty