Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methanol ≥99.9% ACS, Spectrophotometric Grade, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Hydroxy methaneMethyl hydroxideMethyl alcoholMethylolCarbinolWood alcohol

154903-500ML 154903-1L
BJ154903-1LEA 93.27 USD InStock
BJ154903-1L BJ154903-500ML
Methanol ≥99.9% ACS, Spectrophotometric Grade, Riedel-de Haën™
Methanol
  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.

Beilstein Number: 1098229


Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64-65 °C (1.013 hPa)
Melting Pt: -98 °C
Density: 0.79 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/33/12/733312.jpg
Order Now
Specification Test Results
Refractive Index n20/D 1.329 (lit.)
Identity (IR) complying
Assay (GC) Min. 99.90 %
Non-volatile matter Max. 0.001 %
Mixable with H₂O complying
Water (Karl Fischer) Max. 0.1 %
Titrable Acid Max. 0.0003 meq/g
Titrable Base Max. 0.0002 meq/g
KMnO₄ reducing matter complying
Acetaldehyde Max. 0.001 %
Formaldehyde Max. 0.001 %
Acetone (GC) Max. 0.001 %
Reaction against H₂SO₄ complying
Absorbance at 400 nm Max. 0.01
Absorbance 280 nm Max. 0.01
Absorbance at 260 nm Max. 0.04
Absorbance at 240 nm Max. 0.10
Absorbance at 230 nm Max. 0.20
Absorbance at 220 nm Max. 0.40
Absorbance at 210 nm Max. 0.80
Absorbance at 205 nm Max. 1.00
APHA Max. 10
Vapor Pressure 97.68 mmHg ( 20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty