Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

n-Pentane ≥99%, Spectrophotometric Grade, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Pentane

154954-2L
BJ154954-2LEA 220 USD InStock
BJ154954-2L
n-Pentane ≥99%, Spectrophotometric Grade, Riedel-de Haën™
n-Pentane
  • Spectrophotometric grade solvents are high purity solvents made specifically for applications using spectrophotometers.

Beilstein Number: 969132
RTECS Number: RZ9450000
UNSPSC Code: 12191502


Formula: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Boiling Pt: 36-37 °C
Melting Pt: -131 °C
Density: 0.631 g/cm³ (15 °C)
Flash Pt: <-35 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/81/67/738167.jpg
Order Now
Specification Test Results
Identity (IR) complying
Assay (GC) Min. 99.0 %
Refractive index (n 20/D) 1.356 - 1.360
Non-volatile matter Max. 0.0003 %
Water (Karl Fischer) Max. 0.01 %
Absorbance at 400 nm Max. 0.01
Absorbance at 245 nm Max. 0.01
Absorbance at 220 nm Max. 0.06
Absorbance at 210 nm Max. 0.30
Absorbance at 200 nm Max. 0.60
Absorbance at 195 nm Max. 1.00
Vapor Pressure 26.98 psi ( 55 °C)
Vapor Pressure 8.4 psi ( 20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty