Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Petroleum spirit, 60…80 °C, Spectrophotometric Grade, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Petroleum etherGasolinePetrol

261734-1L
BJ261734-1LEA 135.22 USD InStock
BJ261734-1L
Petroleum spirit, 60…80 °C, Spectrophotometric Grade, Riedel-de Haën™
Petroleum spirit
  • Spectrophotometric grade solvents are high purity solvents made specifically for applications using spectrophotometers.

RTECS Number: OI6180000
UNSPSC Code: 12191502


Boiling Pt: 40-60 °C (1.013 hPa)
Density: 0.65 g/cm³ (20 °C)
Flash Pt: -30 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
UN: 3295
ADR: 3,II
Order Now
Specification Test Results
Explosion Limit 8 %
Identity (IR) complying
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.02 %
Absorbance at 400 nm Max. 0.01
Absorbance at 260 nm Max. 0.01
Absorbance at 240 nm Max. 0.05
Absorbance at 230 nm Max. 0.10
Absorbance at 220 nm Max. 0.30
Absorbance at 210 nm Max. 1.0
Vapor Density 2.5 (vs air)
Refractive Index n20/D 1.363 (lit.)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty