Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Carbon disulfide ≥99.9% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur. for IR spectrophotometry, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

31627-1L
BJ31627-1LEA 269.15 USD InStock
BJ31627-1L
Carbon disulfide ≥99.9% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur. for IR spectrophotometry, Riedel-de Haën™
Carbon disulphide
  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.

Beilstein Number: 1098293
RTECS Number: FF6650000
UNSPSC Code: 12190000


Formula: CS₂
MW: 76.14 g/mol
Boiling Pt: 46 °C (1.013 hPa)
Melting Pt: -76 °C
Density: 1.264 g/cm³ (20 °C)
Flash Pt: -30 °C
Storage Temperature: Ambient
MDL Number: MFCD00011321
CAS Number: 75-15-0
EINECS: 200-843-6
UN: 1131
ADR: 3,I
https://us-legacy.vwr.com/stibo/web/std.lang.all/53/94/785394.jpg
Order Now
Specification Test Results
Vapor Density 2.67 (vs air)
Explosion Limit 50 %
Assay (GC) Min. 99.9 %
Boiling range 46 - 47 °C
Density (D 20/4) 1.262 - 1.263
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.01 %
H₂S + foreign org. S-comp. Max. 0.0001 %
Sulfate (SO₄) Max. 0.00025 %
Sulfite (as SO₂) Max. 0.001 %
Benzene Max. 0.001 %
Iodine consuming matters complying
APHA Max. 10
Autoignition Temperature 212 °F

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty