Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Propanol, Semiconductor grade (Honeywell 17829)

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Isopropyl alcoholIsopropyl Rubbing Alcoholsec-Propanolsec-Propyl Alcohol2-Hydroxy propaneDimethyl carbinol2-HydroxypropanePropane, 2-hydroxy-Propan-2-olIPAIsopropanol

40219H-5L
BJ40219H-5LEA 539.72 USD InStock
BJ40219H-5L
2-Propanol, Semiconductor grade (Honeywell 17829)
2-Propanol
We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:

  • Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
  • Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.

Beilstein Number: 635639


Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 77-83 °C (1.013 hPa)
Melting Pt: -90 °C
Density: 0.784-0.786 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/19/69/731969.jpg
Order Now
Specification Test Results
Assay (GC) Min. 99.8 %
Boiling range 81 - 83 °C
Density (D 20/4) 0.785 - 0.787
Refractive index (n 20/D) 1.3770 - 1.3780
Non-volatile matter Max. 5 ppm
Water (Karl Fischer) Max. 500 ppm
Free acid (as C₂H₅COOH) Max. 10 ppm
Silver (Ag) Max. 0.02 ppm
Aluminium (Al) Max. 0.05 ppm
Arsenic (As) Max. 0.01 ppm
Gold (Au) Max. 0.02 ppm
Boron (B) Max. 0.01 ppm
Barium (Ba) Max. 0.1 ppm
Beryllium (Be) Max. 0.01 ppm
Bismuth (Bi) Max. 0.02 ppm
Calcium (Ca) Max. 0.2 ppm
Cadmium (Cd) Max. 0.01 ppm
Cobalt (Co) Max. 0.01 ppm
Chromium (Cr) Max. 0.01 ppm
Copper (Cu) Max. 0.01 ppm
Iron (Fe) Max. 0.1 ppm
Gallium (Ga) Max. 0.02 ppm
Indium (In) Max. 0.02 ppm
Potassium (K) Max. 0.1 ppm
Lithium (Li) Max. 0.02 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.01 ppm
Molybdenum (Mo) Max. 0.01 ppm
Sodium (Na) Max. 0.2 ppm
Nickel (Ni) Max. 0.01 ppm
Lead (Pb) Max. 0.02 ppm
Platinum (Pt) Max. 0.02 ppm
Antimony (Sb) Max. 0.01 ppm
Tin (Sn) Max. 0.02 ppm
Strontium (Sr) Max. 0.02 ppm
Titanium (Ti) Max. 0.01 ppm
Thallium (Tl) Max. 0.02 ppm
Vanadium (V) Max. 0.01 ppm
Zinc (Zn) Max. 0.05 ppm
Zirconium (Zr) Max. 0.01 ppm
Chloride (Cl) Max. 1 ppm
Sulfate (SO₄) Max. 1 ppm
Acetone (GC) Max. 100 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty