Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Methanol VLSI, Semiconductor grade (Honeywell 17744)

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Hydroxy methaneMethyl hydroxideMethyl alcoholMethylolCarbinolWood alcohol

40296H-5L
BJ40296H-5LEA 368.45 USD InStock
BJ40296H-5L
Methanol VLSI, Semiconductor grade (Honeywell 17744)
Methanol
  • comprehensive portfolio for the electronics and semiconductor industry
  • for scientists engaged in education, research, development, prototyping and small scale fabrication of high tech micro- and nanochips
  • Superior purity, stability and exceptional lot-to-lot consistency
  • includes high purity products required for high performance applications such as etchants, solvents or hexamethyldisilazane (HMDS)

Beilstein Number: 1098229


Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64 - 65 °C
Melting Pt: -98 °C
Density: 0.79 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
UN: 1230
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/33/12/733312.jpg
Order Now
Specification Test Results
Particles > 0,5 µm Max. 100 P/ml
Assay (GC) Min. 99.9 %
Spec. resistivity (MOhm·cm) Min. 1.0 MΩ·cm
Water (Karl Fischer) Max. 500 ppm
Free acid (as HCOOH) Max. 20 ppm
Free alkali (as NH₃) Max. 1.0 ppm
Silver (Ag) Max. 0.01 ppm
Aluminium (Al) Max. 0.05 ppm
Arsenic (As) Max. 0.01 ppm
Gold (Au) Max. 0.02 ppm
Boron (B) Max. 0.01 ppm
Barium (Ba) Max. 0.02 ppm
Beryllium (Be) Max. 0.01 ppm
Bismuth (Bi) Max. 0.02 ppm
Calcium (Ca) Max. 0.1 ppm
Cadmium (Cd) Max. 0.01 ppm
Cobalt (Co) Max. 0.01 ppm
Chromium (Cr) Max. 0.01 ppm
Copper (Cu) Max. 0.01 ppm
Iron (Fe) Max. 0.01 ppm
Gallium (Ga) Max. 0.01 ppm
Indium (In) Max. 0.01 ppm
Potassium (K) Max. 0.02 ppm
Lithium (Li) Max. 0.01 ppm
Magnesium (Mg) Max. 0.02 ppm
Manganese (Mn) Max. 0.01 ppm
Molybdenum (Mo) Max. 0.01 ppm
Sodium (Na) Max. 0.1 ppm
Nickel (Ni) Max. 0.01 ppm
Lead (Pb) Max. 0.01 ppm
Platinum (Pt) Max. 0.05 ppm
Antimony (Sb) Max. 0.01 ppm
Tin (Sn) Max. 0.02 ppm
Strontium (Sr) Max. 0.01 ppm
Titanium (Ti) Max. 0.02 ppm
Thallium (Tl) Max. 0.01 ppm
Vanadium (V) Max. 0.01 ppm
Zinc (Zn) Max. 0.02 ppm
Zirconium (Zr) Max. 0.02 ppm
Heavy metals (as Pb) Max. 0.1 ppm
KMnO₄ reducing matter (as O) Max. 2.5 ppm
Aldehydes, ketones (HCHO) Max. 10 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty