Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

2-Propanol, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Reag. USP, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Isopropyl alcoholIsopropyl Rubbing Alcoholsec-Propanolsec-Propyl Alcohol2-Hydroxy propaneDimethyl carbinol2-HydroxypropanePropane, 2-hydroxy-Propan-2-olIPAIsopropanol

72299-4X2.5L 72299-2.5L 72299-6X1L 72299-1L
BJ72299-2.5LEA 94.89 USD InStock
BJ72299-2.5L BJ72299-1L BJ72299-6X1L BJ72299-4X2.5L
2-Propanol, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Reag. USP, Riedel-de Haën™
2-Propanol
ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Beilstein Number: 635639


Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 77-83 °C (1.013 hPa)
Melting Pt: -90 °C
Density: 0.784-0.786 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/19/69/731969.jpg
Order Now
Specification Test Results
Appearance complying
Assay (GC) Min. 99.9 %
IR spectrum complying
Boiling range 82.1 - 82.6 °C
Density (D 20/4) 0.784 - 0.786
Refractive index (n 20/D) 1.3770 - 1.3780
Non-volatile matter Max. 10 ppm
Water (Karl Fischer) Max. 0.1 %
Free acid (as C₂H₅COOH) Max. 0.00074 %
Free alkali (as NH₃) Max. 0.00017 %
Free acid, free alkali Max. 0.0001 meq/g
Peroxides (as H₂O₂) Max. 0.00034 %
Reaction against H₂SO₄ complying
KMnO₄ reducing matter (as O) Max. 5 ppm
Aluminium (Al) Max. 0.5 ppm
Boron (B) Max. 0.02 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.01 ppm
Cobalt (Co) Max. 0.01 ppm
Chromium (Cr) Max. 0.01 ppm
Copper (Cu) Max. 0.01 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.01 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 1 ppm
Nickel (Ni) Max. 0.01 ppm
Lead (Pb) Max. 0.01 ppm
Tin (Sn) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.01 ppm
Heavy metals (as Pb) Max. 1 ppm
Carbonyl compounds (as Acetone) Max. 20 ppm
Carbonyl compounds (as propionaldehyde) Max. 20 ppm
Ethanol (GC) Max. 0.01 %
Methanol (GC) Max. 0.1 %
APHA Max. 10
Solubility in water complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty