Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Pentane ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™

Supplier: Honeywell Research Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Danger
Synonyms: Pentane

76871-1L 76871-2.5L
BJ76871-2.5LEA 416.01 USD InStock
BJ76871-2.5L BJ76871-1L
n-Pentane ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™
n-Pentane
  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.

Beilstein Number: 969132


Formula: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Boiling Pt: 36-37 °C
Melting Pt: -131 °C
Density: 0.631 g/cm³ (15 °C)
Flash Pt: <-35 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/81/67/738167.jpg
Order Now
Specification Test Results
Assay (GC) Min. 99.0 %
Refractive index (n 20/D) 1.357 - 1.359
Non-volatile matter Max. 0.002 %
Water (Karl Fischer) Max. 0.05 %
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
Isopentane (GC) Max. 1.0 %
2-methylpentane (GC) Max. 1.0 %

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty