Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 4 M HCl in water, Fluka™

Supplier: Honeywell Research Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Danger
Synonyms: Muriatic acid

84435-1L
BJ84435-1LEA 66.88 USD InStock
BJ84435-1L
Hydrochloric acid 4 M HCl in water, Fluka™
Hydrochloric acid
Honeywell's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)Buffers available as concentrates or ready-to-useReagents for complexometry:Aminopolycarboxylic acids (EDTA/NTA analogs)
  • Masking agents
  • Indicators for volumetric and complexometric titrations

UNSPSC Code: 12000000


Formula: HCl
MW: 36.46 g/mol
Density: 1.072 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Appearance (color) complying
Appearance (form) complying
Assay 3.95 - 4.10 mol/l

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty