Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Titration solvent ASTM D-4739, LabReady™ Solvent Blend total base number titration solvent, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

WP2162-19P
BJWP2162-19PEA 482.04 USD InStock
BJWP2162-19P
Titration solvent ASTM D-4739, LabReady™ Solvent Blend total base number titration solvent, Burdick & Jackson™
Titration solvent


Boiling Pt: 75,56 °C
Density: 1,0417 g/cm3 (25 °C)
Flash Pt: 18,3 °C
Storage Temperature: Ambient
UN: 1992
ADR: 3,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty