Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Papanicolaou's solution EA 36, VWR® stain for histology, for cytology

Supplier: VWR International
Danger

7042-32
10143-616CS 2957.88 USD InStock
10143-616
Papanicolaou's solution EA 36, VWR® stain for histology, for cytology
Papanicolaou's solution EA 36
Papanicolaou stains are used in conjunction with Hematoxylin nuclear stains in the diagnosis of malignant cytological diseases.

  • Crisp, clear staining
  • Well-defined cytoplasm
  • ready-to-use

Gynecological staining protocols use OG-6 and hematoxylin along with either EA-36 or EA-50. Non gynecological procedures require use of EA-65. EASYpap is a single step cytology staining solution. Combination of EA and OG-6 solutions saves times by providing customer with an easy one step process. FDA approved for in vitro diagnostic use. For Professional Use Only.


Storage Temperature: Ambient
UN: 1170
ADR: 3,II
Order Now
Specification Test Results
Absorbance (Visible Scan 700- 400nm) @626 nm 270 – 350 MA

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty