Would you like to log in again?
Icing Protection Sleeve for Pump Connectors, JULABO
Supplier: Julabo
8970750
89422-318EA 176.49 USD InStock
89422-318
Icing Protection Sleeve for Pump Connectors, JULABO
Tubing Chromatography Tubing
This icing protection sleeve reduces/eliminates ice formation on pump connections during cold operation.
  • Easy installation
This sleeve installs on pump connections to eliminate ice formation during cold temperature operation. The insulated foam installs over pump connections with Velcro.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty