Would you like to log in again?
Nexion ICP-MS Calibration Standard for Set Up 8, VeriSpec®, Ricca Chemical Company
Supplier: RICCA Chemical Company
VeriSpec®
RV010880-100N RV010880-500N
RCRV010880100NEA 367.91 USD InStock
RCRV010880100N RCRV010880500N
Nexion ICP-MS Calibration Standard for Set Up 8, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Multi-Element Standards
8 elements.
1 ppb: Be, Ce, Fe, In, Li, Mg, Pb, U
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty