Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tyloxapol USP

Supplier: BioVision

AB285429-5G AB285429-25G
76335-966EA 711.01 USD InStock
76335-966 76335-968
Tyloxapol USP
Tyloxapol
Tyloxapol is a nonionic liquid polymer of the alkyl aryl polyether alcohol type that is used as a surfactant to aid liquefaction. Tyloxapol is used in the development of non-ionic surfactant-based liposomes (niosomes) for use in drug delivery systems. Tyloxapol may be used to study its mechanism of protecting macrophages from endotoxins.

  • A nonionic liquid surfactant


Storage Temperature: Ultra-Low Temperature Freezer (–80°C)
MDL Number: MFCD00149002
CAS Number: 25301-02-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/35/57/27363557.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty