Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tyloxapol solution 10%

Supplier: BioVision

AB286954-50ML AB286954-250ML
76335-970EA 227.78 USD InStock
76335-970 76335-972
Tyloxapol solution 10%
Tyloxapol
A 10% solution of the nonionic liquid surfactant Tyloxapol (Cat. No.1158) in water

  • A nonionic liquid surfactant


Storage Temperature: Refrigerator
MDL Number: MFCD00149002
CAS Number: 25301-02-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/35/58/27363558.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty