Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

AG-120 (Ivosidenib) ≥98% (by HPLC)

Supplier: BioVision
Synonyms: (2S)-N-(1-(2-chlorophenyl)-2-((3,3-difluorocyclobutyl)amino)-2-oxoethyl)-1-(4-cyanopyridin-2-yl)-N-(5-fluoropyridin-3-yl)-5-oxopyrrolidine-2-carboxamideIvosidenib racemate

AB285377-25MG AB285377-5MG
76335-980EA 699.15 USD InStock
76335-980 76335-978
AG-120 (Ivosidenib) ≥98% (by HPLC)
AG-120 (Ivosidenib)
AG-120 is a potent isocitrate dehydrogenase type 1 (IDH1) inhibitor, with potential antineoplastic activity. Upon oral administration it specifically inhibits a mutated form of IDH1 in the cytoplasm inhibiting the formation of oncometabolite, 2-hydroxyglutarate (2HG). This leads to induction of cellular differentiation and an inhibition of cellular proliferation

  • A potent isocitrate dehydrogenase type 1 (IDH1) inhibitor


Formula: C₂₈H₂₂ClF₃N₆O₆
MW: 582.97 g/mol
Storage Temperature: Freezer
MDL Number: MFCD29044889
CAS Number: 1448347-49-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/35/62/27363562.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty