Would you like to log in again?
ChemMax® 4 Plus Chemical Protective Disposable Coveralls with Respirator-Fit Hood, Double Storm Flap, Attached Boots and Boot Flaps, Lakeland
Supplier: Lakeland Industries
C4T165Y-2X
76359-580CS 1052.36 USD InStock
76359-580
ChemMax® 4 Plus Chemical Protective Disposable Coveralls with Respirator-Fit Hood, Double Storm Flap, Attached Boots and Boot Flaps, Lakeland
Disposable Garments
Chemical protective disposable coverall made from an advanced chemical barrier with heat sealed seams for the highest degeree of chemical protection.
  • Advanced chemical barrier
  • Respirator fit hood
  • Heat sealed seams
  • PermaSURE chemical database
ChemMax® 4 Plus garments are made from an advanced chemical protective barrier to provide the highest level of chemical protection. These garments feature a respirator fit hood, double storm flap, and attached boots with boot flaps for maximum protection.
PermaSURE software provides additional support to all Lakeland ChemMax® 3, ChemMax® 4 Plus, and Interceptor Plus garments by providing a accessable database of more than 4000 chemicals. Calculate safe use times of protective garments, as well as the affects of environmental temperature on those times, via the PermaSURE modeler.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty