Would you like to log in again?
Stainless Steel Bristle and Plywood Scratch Brush, 1×10 Row, Gordon Brush
Supplier: Gordon Brush
11SSPG-12
22940-620EA 68.1 USD InStock
22940-620
Stainless Steel Bristle and Plywood Scratch Brush, 1×10 Row, Gordon Brush
Brushes
Toothbrush-style scratch brush with 0.006" Stainless Steel Bristle and a Plywood Handle in a curved design for knuckle protection.
  • Brush has a 1×10 row configuration
  • Ideal for reaching narrow confned areas
  • Each fill tuft is tied into position by hand
  • Greater bristle density than staple set brushes
The stainless steel bristle, hand-laced with stainless steel wire, remains secure even in wet environments if the wooden handle expands or contracts.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty