Would you like to log in again?
SP Bel-Art H-B Frio-Temp® Bluetooth® Verification Thermometer, Hygrometer, Data Logger, Bel-Art Products, a part of SP
Supplier: Bel-Art
B61011-5100
76305-204CS 1669.06 USD InStock
76305-204
SP Bel-Art H-B Frio-Temp® Bluetooth® Verification Thermometer, Hygrometer, Data Logger, Bel-Art Products, a part of SP
Data Loggers
Enable remote monitoring and alerts by broadcasting readings to any Apple or Android smartphone or device with Bluetooth® 4.0 or better utilizing the free THERMSmart app.
  • Monitor critical environments without a physical connection; maximum signal distance is approximately 40 meters
  • Record temperature data for 30 days
  • Hourly data recorded for current seven days, then maximum and minimum temperatures and humidity recorded for next 23 days
  • Records accurate temperature reading without sample contamination by using a glass bead filled bottle (included) which mimics the sample
  • Alarm sounds to alert user if temperature falls outside of the set parameters
  • App can trigger indicator light for unit identification
  • Individually serialized
Data viewable as a table, graph, or calendar, data from up to six data loggers can be plotted on one graph for direct comparison, also downloadable as a CSV file. Indicators include memory full, active alarm state and low battery, WBGT (Wet Bulb Global Temperature), heat, mold and comfort indices.
Certifications: CE marked. RoHS compliant. Includes Certificate of Compliance indicating accuracy for 4 temperature and 3 humidity points.
Delivery information: Requires 2 AAA batteries (included). Includes multi-language (English, French, German, Italian, Portuguese and Spanish) instructions.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty