Would you like to log in again?
Face Shield Plus, PW96, Portwest
Supplier: Portwest
PW96CLR
76105-172EA 23.01 USD InStock
76105-172
Face Shield Plus, PW96, Portwest
Face Shields
This face shield protects against molten metal splashes and hot solid projections. Superior front and side face protection. Ratchet adjustment for comfort fit.
  • Browguard including clear polycarbonate visor with aluminum edge
  • Protects from impacts, metal splashes and hot solid projections
  • Suitable for a wide range of applications
  • Wheel ratchet size adjustment for easy fitting
  • Simple and secure visor replacement
Replacement visor available (PW99).
Certifications: CE certified. Meet EN 166 (1 BT 3 9), ANSI/ISEA Z87 +
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty